Mucobromic acid, 99+%, Thermo Scientific Chemicals
Mucobromic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mucobromic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 128055000
Price (USD)/ Each
437.00
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
437.00
Add to cart
Mucobromic acid, 99+%, Thermo Scientific Chemicals
Catalog number128055000
Price (USD)/ Each
437.00
-
Add to cart
Chemical Identifiers
CAS488-11-9
IUPAC Name(2Z)-2,3-dibromo-4-oxobut-2-enoate
Molecular FormulaC4HBr2O3
InChI KeyNCNYEGJDGNOYJX-IHWYPQMZSA-M
SMILES[O-]C(=O)C(\Br)=C(\Br)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration with NaOH>=99.0 %
Infrared spectrumConforms
Melting point122°C to 125°C
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only