Thermo Scientific Chemicals

Mucochloric acid, 99%, Thermo Scientific Chemicals

Catalog number: 128065000
500 g, Each
Thermo Scientific Chemicals

Mucochloric acid, 99%, Thermo Scientific Chemicals

Catalog number: 128065000
500 g, Each
Quantity
Catalog number: 128065000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
87-56-9
IUPAC Name
(2Z)-2,3-dichloro-4-oxobut-2-enoic acid
Molecular Formula
C4H2Cl2O3
InChI Key
LUMLZKVIXLWTCI-IHWYPQMZSA-N
SMILES
OC(=O)C(Cl)=C(Cl)C=O
Appearance (Form)
Crystalline powder
Melting point
125°C to 128°C
Water
=<4 % (K.F.)
Appearance (Color)
White to light beige
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text