trans,trans-Muconic acid, 97%, Thermo Scientific Chemicals
trans,trans-Muconic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans,trans-Muconic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 128070050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3588-17-8
IUPAC Name(2E,4E)-hexa-2,4-dienedioic acid
Molecular FormulaC6H6O4
InChI KeyTXXHDPDFNKHHGW-ZPUQHVIOSA-N
SMILESOC(=O)C=CC=CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only