Thermo Scientific Chemicals

Myrcene, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 128085000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Myrcene, 90%, tech., stabilized, Thermo Scientific Chemicals

Catalog number: 128085000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 128085000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123-35-3
IUPAC Name
7-methyl-3-methylideneocta-1,6-diene
Molecular Formula
C10H16
InChI Key
UAHWPYUMFXYFJY-UHFFFAOYSA-N
SMILES
CC(C)=CCCC(=C)C=C
Appearance (Form)
Liquid
Refractive index
1.4660 to 1.4720 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
GC
>=88 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text