Thermo Scientific Chemicals

alpha-Naphthoflavone, 97%, Thermo Scientific Chemicals

Catalog number: 128160010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

alpha-Naphthoflavone, 97%, Thermo Scientific Chemicals

Catalog number: 128160010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 128160010
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
604-59-1
IUPAC Name
2-phenyl-4H-benzo[h]chromen-4-one
Molecular Formula
C19H12O2
InChI Key
VFMMPHCGEFXGIP-UHFFFAOYSA-N
SMILES
O=C1C=C(OC2=C1C=CC1=CC=CC=C21)C1=CC=CC=C1
Appearance (Form)
Crystalline powder or crystals
HPLC
>=99.50 %
Appearance (Color)
Off-white to light yellow
Melting point
155°C to 158°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text