Thermo Scientific Chemicals

1-Naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 128180250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

1-Naphthoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 128180250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 128180250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-55-5
IUPAC Name
naphthalene-1-carboxylic acid
Molecular Formula
C11H8O2
InChI Key
LNETULKMXZVUST-UHFFFAOYSA-N
SMILES
OC(=O)C1=C2C=CC=CC2=CC=C1
HPLC
>=97.5 %
Appearance (Form)
Powder
Appearance (Color)
Light yellow to brown
Melting point
160°C to 164°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text