1-Naphthol, 99+%
1-Naphthol, 99+%
1-Naphthol, 99+%
Thermo Scientific Chemicals

1-Naphthol, 99+%

1-Naphthol, 99+%, C10H8O, CAS Number-90-15-3, 1-hydroxynaphthalene, 1-naphthalenol, 1-naphthol, alpha-naphthol, basf ursol ern, fouramine ern, fourrine ern, furro er, naphthol, tertral ern, 100g, 06, 596, 278.0 deg.C to 280.0 deg.C, 100.0, 99.0, CHEBI:10319, Beige-Gray to Brown, 201-969-4, 125 deg.C | CAS: 90-15-3 | C10H8O | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
500 g
Packaging:
View all
Glass bottle
Catalog number 128191000
Price (USD)
71.65
Online Exclusive
79.20
Save 7.55 (10%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Online Exclusive
79.20
Save 7.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS90-15-3
IUPAC Namenaphthalen-1-ol
Molecular FormulaC10H8O
InChI KeyKJCVRFUGPWSIIH-UHFFFAOYSA-N
SMILESOC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point95°C to 97°C
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Beige to grey to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link