Thermo Scientific Chemicals

Neopentyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 128250100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Neopentyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 128250100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 128250100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-84-3
IUPAC Name
2,2-dimethylpropan-1-ol
Molecular Formula
C5H12O
InChI Key
KPSSIOMAKSHJJG-UHFFFAOYSA-N
SMILES
CC(C)(C)CO
Appearance (Form)
Waxy crystalline solid
GC
>=98.5 %
Appearance (Color)
Colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text