Nickel acetylacetonate, 96%, Thermo Scientific Chemicals
Nickel acetylacetonate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel acetylacetonate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 128260250
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
Add to cart
Nickel acetylacetonate, 96%, Thermo Scientific Chemicals
Catalog number128260250
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3264-82-2
IUPAC Namenickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14NiO4
InChI KeyBMGNSKKZFQMGDH-FDGPNNRMSA-L
SMILES[Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=95 %
Appearance (Color)Green
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only