Thermo Scientific Chemicals

Nicotinamide, 99%

Catalog number: 128271000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Nicotinamide, 99%

Catalog number: 128271000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 128271000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-92-0
IUPAC Name
pyridine-3-carboxamide
Molecular Formula
C6H6N2O
InChI Key
DFPAKSUCGFBDDF-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CN=C1
Appearance (Form)
Crystalline powder or crystals
Loss on drying
=<0.5 % (4 h) (silica gel)
pH
6.0 to 7.5 (5 % in water at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text