Nipecotamide, 95%, Thermo Scientific Chemicals
Nipecotamide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nipecotamide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
Glass Bottle
Catalog number 128300500
Price (USD)/ Each
622.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
622.00
Add to cart
Nipecotamide, 95%, Thermo Scientific Chemicals
Catalog number128300500
Price (USD)/ Each
622.00
-
Add to cart
Chemical Identifiers
CAS4138-26-5
IUPAC Name(3R)-3-carbamoylpiperidin-1-ium
Molecular FormulaC6H13N2O
InChI KeyBVOCPVIXARZNQN-RXMQYKEDSA-O
SMILESNC(=O)[C@@H]1CCC[NH2+]C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=94 %
Infrared spectrumConforms
Appearance (Color)Yellow
Melting point102°C to 108°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only