o-Nitroacetophenone, 95%, Thermo Scientific Chemicals
o-Nitroacetophenone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Nitroacetophenone, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
20 g
2.5 g
Packaging:
Glass bottle
Catalog number 128320200
Price (USD)/ Each
132.00
-
Add to cart
Quantity:
20 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
132.00
Add to cart
o-Nitroacetophenone, 95%, Thermo Scientific Chemicals
Catalog number128320200
Price (USD)/ Each
132.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-(2-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeySUGXZLKUDLDTKX-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1[N+]([O-])=O
Molecular Weight (g/mol)165.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
Infrared spectrumConforms
Appearance (Form)Low melting solid
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only