Thermo Scientific Chemicals

m-Nitroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 128335000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

m-Nitroacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: 128335000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 128335000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-89-1
IUPAC Name
1-(3-nitrophenyl)ethan-1-one
Molecular Formula
C8H7NO3
InChI Key
ARKIFHPFTHVKDT-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Form)
Crystalline powder or crystals and/or chunks
Appearance (Color)
Yellow to yellow-green to yellow-brown
Infrared spectrum
Conforms
GC
>=97.5 %
Melting point
76°C to 80°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text