Thermo Scientific Chemicals

3-Nitrobenzoic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 128451000
100 g, Each
Thermo Scientific Chemicals

3-Nitrobenzoic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 128451000
100 g, Each
Quantity
Catalog number: 128451000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-92-6
IUPAC Name
3-nitrobenzoic acid
Molecular Formula
C7H5NO4
InChI Key
AFPHTEQTJZKQAQ-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
HPLC
>=98.5 %
Appearance (Color)
White to light yellow or light green
Melting point
139°C to 143°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text