4-Nitrobenzoic acid, 99+%, Thermo Scientific Chemicals
4-Nitrobenzoic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrobenzoic acid, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
100 g
Packaging:
Plastic bottle
Catalog number 128460010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS62-23-7
IUPAC Name4-nitrobenzoic acid
Molecular FormulaC7H5NO4
InChI KeyOTLNPYWUJOZPPA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point238°C to 242°C
Appearance (Form)Powder or crystalline powder or crystals
Titration with NaOH99.0 to 101.0 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only