2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals
2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Catalog number 128510250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS612-25-9
IUPAC Name(2-nitrophenyl)methanol
Molecular FormulaC7H7NO3
InChI KeyBWRBVBFLFQKBPT-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Beige to light brown
Melting point69°C to 73°C
Appearance (Form)Crystalline powder
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only