2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals
2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass Bottle
Catalog number 128511000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2-nitrophenyl)methanol
Molecular FormulaC7H7NO3
InChI KeyBWRBVBFLFQKBPT-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1[N+]([O-])=O
Molecular Weight (g/mol)153.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to light brown
Melting point69°C to 73°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
1x1 image pixel for data collection