Thermo Scientific Chemicals

4-(4-Nitrobenzyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 128550500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(4-Nitrobenzyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: 128550500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 128550500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1083-48-3
IUPAC Name
4-[(4-nitrophenyl)methyl]pyridine
Molecular Formula
C12H10N2O2
InChI Key
MNHKUCBXXMFQDM-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(CC2=CC=NC=C2)C=C1
Appearance (Color)
Light yellow
Melting point
69°C to 72°C
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text