Nitro Blue Tetrazolium Chloride monohydrate, 90%
Nitro Blue Tetrazolium Chloride monohydrate, 90%
Thermo Scientific Chemicals

Nitro Blue Tetrazolium Chloride monohydrate, 90%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 128590050
Price (USD)/ Each
679.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
799.00 
Save 119.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
679.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
799.00 
Save 119.35 (15%)
Add to cart
Nitro Blue Tetrazolium Chloride monohydrate, 90%
Catalog number128590050
Price (USD)/ Each
679.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
799.00 
Save 119.35 (15%)
-
Add to cart
Chemical Identifiers
CAS298-83-9, 67-56-1, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
Appearance (Form)Powder
HPLC>=88 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only