5-Nitroindole, 99%, Thermo Scientific Chemicals
5-Nitroindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitroindole, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 128700050
Price (USD)/ Each
80.60
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
80.60
Add to cart
5-Nitroindole, 99%, Thermo Scientific Chemicals
Catalog number128700050
Price (USD)/ Each
80.60
-
Add to cart
Chemical Identifiers
CAS6146-52-7
IUPAC Name5-nitro-1H-indole
Molecular FormulaC8H6N2O2
InChI KeyOZFPSOBLQZPIAV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder or needles
GC>=98.5 %
Melting point140°C to 143°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only