5-Nitroisatin, 97%, Thermo Scientific Chemicals
5-Nitroisatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Nitroisatin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 128720250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name5-nitro-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC8H4N2O4
InChI KeyUNMYHYODJHKLOC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC(=O)C(=O)C2=C1
Molecular Weight (g/mol)192.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only