Thermo Scientific Chemicals

2-Nitrophenol, 99%, Thermo Scientific Chemicals

Catalog number: 128745000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Nitrophenol, 99%, Thermo Scientific Chemicals

Catalog number: 128745000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 128745000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
88-75-5
IUPAC Name
2-nitrophenol
Molecular Formula
C6H5NO3
InChI Key
IQUPABOKLQSFBK-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1[N+]([O-])=O
Appearance (Color)
Yellow to yellow-green
Appearance (Form)
crystalline mass
Melting point
43°C to 47°C
Appearance (Form)
Powder or crystals and/or chunks or low melting
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text