4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 128750050
Price (USD)/ Each
31.70
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
31.70
Add to cart
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Catalog number128750050
Price (USD)/ Each
31.70
-
Add to cart
Chemical Identifiers
CAS830-03-5
IUPAC Name4-nitrophenyl acetate
Molecular FormulaC8H7NO4
InChI KeyQAUUDNIGJSLPSX-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point76°C to 79°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only