Thermo Scientific Chemicals

4-Nitrophenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 128760500
50 g, Each
Thermo Scientific Chemicals

4-Nitrophenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog number: 128760500
50 g, Each
Quantity
Catalog number: 128760500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-03-0
IUPAC Name
2-(4-nitrophenyl)acetic acid
Molecular Formula
C8H7NO4
InChI Key
YBADLXQNJCMBKR-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
Yellow to beige
Melting point
153°C to 156°C
Titration with NaOH
>=98.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text