4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 128860100
Price (USD)/ Each
328.65
Online exclusive
386.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
328.65
Online exclusive
386.00 
Save 57.35 (15%)
Add to cart
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
Catalog number128860100
Price (USD)/ Each
328.65
Online exclusive
386.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium hexahydrate 4-nitrophenyl phosphate
Molecular FormulaC6H16NNa2O12P
InChI KeyKAKKHKRHCKCAGH-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
Molecular Weight (g/mol)371.14
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.0 to 102.0 %
HPLC=<0.1 % free 4-nitrophenol
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water27.5 to 31.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only