4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, Thermo Scientific Chemicals

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 128860500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS333338-18-4
IUPAC Namedisodium hexahydrate 4-nitrophenyl phosphate
Molecular FormulaC6H16NNa2O12P
InChI KeyKAKKHKRHCKCAGH-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO498.0 to 102.0 %
HPLC=<0.1 % free 4-nitrophenol
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water27.5 to 31.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only