4-Nitropyridine N-oxide, 97%, Thermo Scientific Chemicals
4-Nitropyridine N-oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitropyridine N-oxide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 128920050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1124-33-0
IUPAC Name4-nitropyridin-1-ium-1-olate
Molecular FormulaC5H4N2O3
InChI KeyRXKNNAKAVAHBNK-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=[N+]([O-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder or crystals
Melting point159°C to 164°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only