2-Nitrotoluene, 99+%, Thermo Scientific Chemicals
2-Nitrotoluene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Nitrotoluene, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 129030010
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.90 
Save 11.25 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.90 
Save 11.25 (14%)
Add to cart
2-Nitrotoluene, 99+%, Thermo Scientific Chemicals
Catalog number129030010
Price (USD)/ Each
66.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
77.90 
Save 11.25 (14%)
-
Add to cart
Chemical Identifiers
CAS88-72-2
IUPAC Name1-methyl-2-nitrobenzene
Molecular FormulaC7H7NO2
InChI KeyPLAZTCDQAHEYBI-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to yellow-green
Infrared spectrumConforms
Refractive index1.5445 to 1.5465 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only