Thermo Scientific Chemicals

2-Nitrotoluene, 99+%, Thermo Scientific Chemicals

Catalog number: 129030025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Nitrotoluene, 99+%, Thermo Scientific Chemicals

Catalog number: 129030025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 129030025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-72-2
IUPAC Name
1-methyl-2-nitrobenzene
Molecular Formula
C7H7NO2
InChI Key
PLAZTCDQAHEYBI-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1[N+]([O-])=O
Infrared spectrum
Conforms
Refractive index
1.5445 to 1.5465 (20°C, 589 nm)
Appearance (Color)
Clear yellow to yellow-green
Appearance (Form)
Liquid
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text