4-Nitrotoluene, 99%, Thermo Scientific Chemicals
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrotoluene, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number 129051000
Price (USD)/ Each
25.65
Online exclusive
28.70 
Save 3.05 (11%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.65
Online exclusive
28.70 
Save 3.05 (11%)
Add to cart
4-Nitrotoluene, 99%, Thermo Scientific Chemicals
Catalog number129051000
Price (USD)/ Each
25.65
Online exclusive
28.70 
Save 3.05 (11%)
-
Add to cart
Chemical Identifiers
CAS99-99-0
IUPAC Name1-methyl-4-nitrobenzene
Molecular FormulaC7H7NO2
InChI KeyZPTVNYMJQHSSEA-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
GC>=98.5 %
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point51°C to 54°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only