Norbornylene, 99%, stabilized, Thermo Scientific Chemicals
Norbornylene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Norbornylene, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Glass bottle
Catalog number 129240250
Price (USD)/ Each
25.90
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
25.90
Add to cart
Norbornylene, 99%, stabilized, Thermo Scientific Chemicals
Catalog number129240250
Price (USD)/ Each
25.90
-
Add to cart
Chemical Identifiers
CAS498-66-8
IUPAC Namebicyclo[2.2.1]hept-2-ene
Molecular FormulaC7H10
InChI KeyJFNLZVQOOSMTJK-UHFFFAOYNA-N
SMILESC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer0.1 % phenolic antioxidant
Appearance (Color)White
Infrared spectrumConforms
Melting point42°C to 46°C
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only