Thermo Scientific Chemicals

Norcamphor, 99%, Thermo Scientific Chemicals

Catalog number: 129251000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Norcamphor, 99%, Thermo Scientific Chemicals

Catalog number: 129251000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 129251000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
497-38-1
IUPAC Name
bicyclo[2.2.1]heptan-2-one
Molecular Formula
C7H10O
InChI Key
KPMKEVXVVHNIEY-UHFFFAOYNA-N
SMILES
O=C1CC2CCC1C2
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Color)
Colorless to white
Appearance (Form)
Adhering crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text