Thermo Scientific Chemicals

Norharman, 98%, Thermo Scientific Chemicals

Catalog number: 129261000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

Norharman, 98%, Thermo Scientific Chemicals

Catalog number: 129261000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 129261000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
244-63-3
IUPAC Name
9H-pyrido[3,4-b]indole
Molecular Formula
C11H8N2
InChI Key
AIFRHYZBTHREPW-UHFFFAOYSA-N
SMILES
N1C2=C(C=CC=C2)C2=C1C=NC=C2
Appearance (Color)
White to yellow to light beige
Appearance (Form)
Powder
Melting point
198°C to 202°C
Titration Tertiary base
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text