Thermo Scientific Chemicals

Octacosane, 99+%, Thermo Scientific Chemicals

Catalog number: 129271000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Octacosane, 99+%, Thermo Scientific Chemicals

Catalog number: 129271000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 129271000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
630-02-4
Appearance (Form)
Glistening powder or flakes
Infrared spectrum
Conforms
GC
>=99.0 %
Melting point
61°C to 64°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text