Thermo Scientific Chemicals

Octadecane, 99+%, Thermo Scientific Chemicals

Catalog number: 129290010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Octadecane, 99+%, Thermo Scientific Chemicals

Catalog number: 129290010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 129290010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
593-45-3
IUPAC Name
octadecane
Molecular Formula
C18H38
InChI Key
RZJRJXONCZWCBN-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC
GC
>=99.0 %
Appearance (Form)
Solid or liquid
Infrared spectrum
Conforms
Melting point
28°C to 31°C
Appearance (Color)
White to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text