1-Octadecanol, 95%, Thermo Scientific Chemicals
1-Octadecanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octadecanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 129300010
Price (USD)/ Each
51.65
Online exclusive
60.70 
Save 9.05 (15%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
60.70 
Save 9.05 (15%)
Add to cart
1-Octadecanol, 95%, Thermo Scientific Chemicals
Catalog number129300010
Price (USD)/ Each
51.65
Online exclusive
60.70 
Save 9.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoctadecan-1-ol
Molecular FormulaC18H38O
InChI KeyGLDOVTGHNKAZLK-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol)270.50
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or flakes or pellets
Infrared spectrumConforms
GC>=94 %
Melting point55°C to 60°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only