1-Octadecene, 90%, tech., Thermo Scientific Chemicals
1-Octadecene, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Octadecene, 90%, tech., Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number 129310025
Price (USD)/ Each
83.65
Online exclusive
92.70 
Save 9.05 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.70 
Save 9.05 (10%)
Add to cart
1-Octadecene, 90%, tech., Thermo Scientific Chemicals
Catalog number129310025
Price (USD)/ Each
83.65
Online exclusive
92.70 
Save 9.05 (10%)
-
Add to cart
Chemical Identifiers
CAS112-88-9
IUPAC Nameoctadec-1-ene
Molecular FormulaC18H36
InChI KeyCCCMONHAUSKTEQ-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
Refractive index1.4435 to 1.4455 (20°C, 589 nm)
Infrared spectrumConforms
GC>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only