n-Octane, 99+%, extra pure, Thermo Scientific Chemicals
n-Octane, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Octane, 99+%, extra pure, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 129375000
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
Add to cart
n-Octane, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number129375000
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS111-65-9
IUPAC Nameoctane
Molecular FormulaC8H18
InChI KeyTVMXDCGIABBOFY-UHFFFAOYSA-N
SMILESCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Density(15°C) 0.706 to 0.710 kg/l
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only