1,8-Octanediol, 98%, Thermo Scientific Chemicals
1,8-Octanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,8-Octanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 129380250
Price (USD)/ Each
40.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
40.30
Add to cart
1,8-Octanediol, 98%, Thermo Scientific Chemicals
Catalog number129380250
Price (USD)/ Each
40.30
-
Add to cart
Chemical Identifiers
CAS629-41-4
IUPAC Nameoctane-1,8-diol
Molecular FormulaC8H18O2
InChI KeyOEIJHBUUFURJLI-UHFFFAOYSA-N
SMILESOCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Colorless to white
Water=<0.5 % (K.F.)
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only