Thermo Scientific Chemicals

Octanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 129390010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Octanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 129390010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 129390010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-07-2
IUPAC Name
octanoic acid
Molecular Formula
C8H16O2
InChI Key
WWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILES
CCCCCCCC(O)=O
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.4260 to 1.4300 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text