Octanoic acid, 99%
Octanoic acid, 99%
Octanoic acid, 99%
Thermo Scientific Chemicals

Octanoic acid, 99%

Octanoic acid, 99%, C8H16O2, CAS Number-124-07-2, 1-heptanecarboxylic acid, caprylic acid, enantic acid, n-caprylic acid, neo-fat 8, n-octanoic acid, n-octoic acid, n-octylic acid, octoic acid, octylic acid, 1L, 385 to 393mg KOH/g, 98.5% min. (GC), 02, 347, 238.0 deg.C, 100.0, CHEBI:28837 | CAS: 124-07-2 | C8H16O2 | 144.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 129390025
Price (USD)
82.65
Special offer
Online exclusive
Ends: 26-Sep-2025
101.00
Save 18.35 (18%)
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
82.65
Special offer
Online exclusive
Ends: 26-Sep-2025
101.00
Save 18.35 (18%)
Each
Add to cart
Chemical Identifiers
CAS124-07-2
IUPAC Nameoctanoic acid
Molecular FormulaC8H16O2
InChI KeyWWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILESCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4260 to 1.4300 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link