Thermo Scientific Chemicals

gamma-Octanoic lactone, 98%, Thermo Scientific Chemicals

Catalog number: 129400250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

gamma-Octanoic lactone, 98%, Thermo Scientific Chemicals

Catalog number: 129400250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 129400250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104-50-7
IUPAC Name
5-butyloxolan-2-one
Molecular Formula
C8H14O2
InChI Key
IPBFYZQJXZJBFQ-UHFFFAOYNA-N
SMILES
CCCCC1CCC(=O)O1
GC
>=97.5 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.4420 to 1.4470 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text