tert-Octylamine, 95%, Thermo Scientific Chemicals
tert-Octylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Octylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number 129505000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-45-9
IUPAC Name2,4,4-trimethylpentan-2-amine
Molecular FormulaC8H19N
InChI KeyQIJIUJYANDSEKG-UHFFFAOYSA-N
SMILESCC(C)(C)CC(C)(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Refractive index1.4235 to 1.4255 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<25 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only