Thermo Scientific Chemicals

Orcinol Monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 129550250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Orcinol Monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 129550250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 129550250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6153-39-5
IUPAC Name
5-methylbenzene-1,3-diol hydrate
Molecular Formula
C7H10O3
InChI Key
NBKPNAMTHBIMLA-UHFFFAOYSA-N
SMILES
O.CC1=CC(O)=CC(O)=C1
Appearance (Color)
White to beige to pink-brown
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
56°C to 61°C
Water
10 to 14 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text