Orotic acid monohydrate, 98%, Thermo Scientific Chemicals
Orotic acid monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Orotic acid monohydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 129571000
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.40 
Save 7.75 (14%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.40 
Save 7.75 (14%)
Add to cart
Orotic acid monohydrate, 98%, Thermo Scientific Chemicals
Catalog number129571000
Price (USD)/ Each
47.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
55.40 
Save 7.75 (14%)
-
Add to cart
Chemical Identifiers
CAS50887-69-9
IUPAC Name2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid
Molecular FormulaC5H4N2O4
InChI KeyPXQPEWDEAKTCGB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=O)NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Loss on drying=<12.0 % (1 g, 130°C, 3 hrs)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only