Thermo Scientific Chemicals

Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 129600010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 129600010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 129600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6153-56-6
IUPAC Name
oxalic acid dihydrate
Molecular Formula
C2H6O6
InChI Key
GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES
O.O.OC(=O)C(O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
98°C to 102°C
Titration with KMnO4
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text