Thermo Scientific Chemicals

Oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 129610025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 129610025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 129610025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-37-8
IUPAC Name
oxalic dichloride
Molecular Formula
C2Cl2O2
InChI Key
CTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILES
ClC(=O)C(Cl)=O
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4285 to 1.4305 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text