Oxalyl chloride, 98%, Thermo Scientific Chemicals
Oxalyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxalyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 129610250
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
Add to cart
Oxalyl chloride, 98%, Thermo Scientific Chemicals
Catalog number129610250
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
CAS79-37-8
IUPAC Nameoxalic dichloride
Molecular FormulaC2Cl2O2
InChI KeyCTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILESClC(=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4285 to 1.4305 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only