Oxalyl chloride, 98%
Oxalyl chloride, 98%
Oxalyl chloride, 98%
Oxalyl chloride, 98%
Thermo Scientific Chemicals

Oxalyl chloride, 98%

CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 129611000
Price (USD)
89.40
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
89.40
Each
Add to cart
Chemical Identifiers
CAS79-37-8
IUPAC Nameoxalic dichloride
Molecular FormulaC2Cl2O2
InChI KeyCTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILESClC(=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4285 to 1.4305 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471971Certificate of AnalysisMay 17, 2025129610025, 129611000, 129610250, 129610010
A0468321Certificate of AnalysisJan 31, 2025129610025, 129611000, 129610250, 129610010
A0465447Certificate of AnalysisAug 14, 2024129610025, 129611000, 129610250, 129610010
A0438812Certificate of AnalysisAug 25, 2023129610025, 129611000, 129610250, 129610010
A0430429Certificate of AnalysisMay 02, 2023129610010, 129610025, 129610250, 129611000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection