Thermo Scientific Chemicals

Palmitic acid, 98%

Catalog number: 129702500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Palmitic acid, 98%

Catalog number: 129702500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 129702500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-10-3
IUPAC Name
hexadecanoic acid
Molecular Formula
C16H32O2
InChI Key
IPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC(O)=O
Appearance (Form)
Powder or granules or flakes
GC
>=97.5 %
Saponification value
216 to 224 mg KOH/g
Infrared spectrum
Conforms
Acid value
215 to 233 mg KOH/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text